missing translation for 'onlineSavingsMsg'
Learn More

Acrylonitrile(stabilized with 35-45 ppm 4-hydroxy anisole (H750015)), TRC

Product Code. 30310151
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310151 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310151 Supplier Toronto Research Chemicals Supplier No. TRCA19137010ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylonitrile
CAS 107-13-1
Purity Grade Notes GC
Molecular Formula C3 H3 N
Quantity 10 mL
Synonym Acrylonitrile (8CI), Acrylon, Carbacryl, Cyanoethene, Cyanoethylene, Fumigrain, NSC 6362, Propenenitrile, VCN, Ventox, Vinyl cyanide
InChI Formula InChI=1S/C3H3N/c1-2-3-4/h2H,1H2
SMILES C=CC#N
IUPAC Name prop-2-enenitrile
Molecular Weight (g/mol) 53.06
Formula Weight 53.0265
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.