missing translation for 'onlineSavingsMsg'
Learn More

Actein, MedChemExpress

Product Code. 16401768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401768 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401768 Supplier Medchem Express Supplier No. HYN6872
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Actein is a triterpene glycoside isolated from the rhizomes of Cimicifuga foetida. Actein suppresses cell proliferation, induces autophagy and apoptosis through promoting ROS/JNK activation, and blunting AKT pathway in human bladder cancer. Actein has little toxicity in vivo.

Actein is a triterpene glycoside isolated from the rhizomes of Cimicifuga foetida. Actein suppresses cell proliferation, induces autophagy and apoptosis through promoting ROS/JNK activation, and blunting AKT pathway in human bladder cancer. Actein has little toxicity in vivo.

Specifications

Chemical Name or Material Actein
CAS 18642-44-9
Color White
Purity Grade Notes Research
Molecular Formula C37H56O11
Quantity 5 mg
SMILES C[C@]12[C@@]3([H])[C@@]4(C[C@@H](OC(C)=O)[C@@]1([C@]5([H])[C@](O[C@]6([C@H]7[C@]([C@@H](O)O6)(C)O7)C[C@H]5C)([H])C2)C)[C@@]8([C@@](C(C)([C@@H](O[C@@]9([H])[C@@H]([C@H]([C@H](O)CO9)O)O)CC8)C)([H])CC3)C4
Molecular Weight (g/mol) 676.83
Formula Weight 676.83
Percent Purity 98.58%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.