Learn More
Ecolab™ Acti - Chlor Plus Tablet
Product Code. 15719854
Change view
Click to view available options
Quantity:
150 Tablets
Unit Size:
Pack of 6
Product Code. Quantity Unit Size
15719854 150 Tablets Pack of 6
1 options
Product Code. 15719854 Supplier Ecolab™ Supplier No. 3077170
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for the disinfection of catering equipment, blood spills, the general clinical environment, babies' bottles and sinks and drains
  • Actichlor disinfectant products are effective against all bacteria, yeasts, spores and viruses including HIV and Hepatitis B
Specifications
Concentration 1.7 g
Quantity 150 Tablets