missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 27-905 Nitrile-Coated Gloves with Ansell Grip™ Technology

Product Code. 16895190
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16895190 11 12 pairs
16865190 8 12 pairs
16875190 9 12 pairs
16885190 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16895190 Supplier Ansell Supplier No. 27905110
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

New and improved nitrile coating is flexible and comfortable, while also providing mechanical protection. Ansell Grip™ Technology requires less force on slippery objects, reduces hand fatigue, and increases productivity.

  • Ideal for tough applications with a combination of three technologies: Ansell Grip™ Technology, Fortix™ Technology and Ripel™ Technology
  • Ansell's unique nitrile formulation is now more durable with patented Fortix™ Technology to offer resistance to abrasive and sharp materials
  • Ansell Grip™ Technology enhances grip in dry, wet, and oily environments
  • Full coating with Ripel™ Technology provides an outstanding barrier to grease and oil, without degrading like leather or cotton
  • Soft polycotton liner provides a cooler feeling to the hands
  • Safety cuff extends protection up the wrist
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Polycotton, Nitrile
Product Type Heavy-duty Nitrile Gloves
Size 11
EN388 Standard 4221C
For Use With (Application) Machinery, Equipment Operation, Oil Drilling, and Maintenance
Glove Cuff Color Natural
Glove Cuff Style Safety
Liner Material Polycotton
Series 27-905
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.