missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves

Product Code. p-7215005
Change view
Click to view available options
Size:
7
9
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16805250 7 12 pairs
16815250 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16805250 Supplier Ansell Supplier No. 78102070
Add to basket
Add to basket
This item is not returnable. View return policy

Seamless knit gloves with a thermal acrylic liner to provide comfort and warmth in cold or refrigerated work environments.

  • Ideal for keeping hands warm in food processing and service applications
  • Seamless knit, thermal acrylic liner provides comfort and warmth in cold or refrigerated work environments
  • Uncoated liner can be used alone, or as an underglove
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant
Coating Material Uncoated
Color Blue
Exterior Finish No Texture
Hand Specific
Material Acrylic
Product Type Thermal Mechanic's Gloves
Size 7
EN388 Standard 113XB
For Use With (Application) Food Processing
Liner Material Acrylic
Series 78-102
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.