missing translation for 'onlineSavingsMsg'
Learn More

ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm

Product Code. 16825250
Change view
Click to view available options
Size:
7
9
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16825250 7 12 pairs
16835250 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16825250 Supplier ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm Supplier No. 78202070
  • ONLINE EXCLUSIVE PRICE
    111.70 €
    74.46 € / 12 pairs
    Save 37.24 €
    33% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Seamless knit gloves with a thermal acrylic liner to provide comfort and warmth in cold or refrigerated work environments. Micro-dot coated palm increases abrasion resistance and enhances grip capabilities.

  • Ideal for keeping hands warm in food processing and service applications
  • Seamless knit, thermal acrylic liner provides comfort and warmth in cold or refrigerated work environments
  • Uncoated liner can be used alone, or as an underglove
  • Micro-dot coating on the palm enhances grip capabilities
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant
Coating Material PVC
Color Blue
Exterior Finish No Texture
Hand Specific
Material Acrylic
Product Type Thermal Mechanic's Gloves
Size 7
EN388 Standard 312XB
For Use With (Application) Food Processing
Liner Material Acrylic
Series 78-202
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.