missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 97-012 General Purpose Hi-Viz™ Gloves

Product Code. 16845260 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16845260 10 12 pairs
16815260 7 12 pairs
16825260 8 12 pairs
16835260 9 12 pairs
16855260 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16845260 Supplier Ansell Supplier No. 97012100
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are engineered to combine all-day comfort, dexterity, enhanced grip, and protection. Specially designed to keep workers safe in low-light or low-visibility environments.

  • High visibility on back of the hand and palm side increases safety
  • 15-gauge liner minimizes perspiration and offers comfortable all day wear
  • Ergonomically designed forms provide naturally curved, relaxed fit to reduce hand fatigue
  • Provides dexterity for the ability to pick up small items
  • Enhances grip in wet and dry conditions
  • Silicone free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A1
Hand Specific
Latex Free Yes
Material Nylon
Product Type Multi-activity Gloves
Size 10
EN388 Standard 3131A
Series 97-012
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.