missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ ActivArmr™ Workguard™ 43-216 Series Yellow Aramid Fiber Heavy Weight Gloves

Product Code. 12679721 Shop All Ansell Products
Change view
Click to view available options
Size:
10
Unit Size:
6 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12679721 10 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12679721 Supplier Ansell Edmont™ Supplier No. 43216100
  • ONLINE EXCLUSIVE PRICE
    121.00 €
    77.41 € / 6 pairs
    Save 43.59 €
    36% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heavy-duty special purpose glove offering high levels of molten splash resistance.

  • Heavy duty glove
  • Ideal for many different applications requiring thermal protection
  • Provides superior durability and good protection from cuts, punctures and abrasions
  • Certified as Category III glove for protection against high temperatures and is rated with the EN 407 performance levels 413X4X
  • Suitable for large number of applications requiring heat resistance, including welding, operating machinery and working in environments with risk of heat or molten splash
  • With soft inner jersey palm lining and cotton denim cuff is comfortable to wear
  • Made of tough-wearing leather heavy-duty glove is designed with palm reinforcement for added abrasion resistance
  • Seams are made in Kevlar™ Aramid fibers and are reinforced by pieces of cowhide, this construction provides superior mechanical protection
  • Durability and mechanical protection

Applications:

  • Welding
  • Loading and unloading with risk of heat or molten splash
TRUSTED_SUSTAINABILITY

Specifications

Back Material Dupont™ Kevlar™ aramid fibers
Cuff Material Dupont™ Kevlar™ aramid fibers
Accelerator Free Yes
Applications Industrial
Coating Material Neoprene
Color Yellow
Latex Free Yes
Material Cowhide Leather
Product Type Heavyweight Gloves
Size 10
Glove Cuff Color Yellow
Glove Cuff Style Gauntlet
Liner Material Cotton Jersey
Lining Lined
Quantity 72 Pairs
Resistant To Heat, cuts, tears and abrasion resistance
Series 43-216
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.