missing translation for 'onlineSavingsMsg'
Learn More

Edwards Vacuum™ Activated Alumina

Product Code. 12935244
Click to view available options
Capacity (Metric):
0.45 kg
Unit Size:
450g
This item is not returnable. View return policy

Product Code. 12935244

Brand: Edwards Vacuum™ H02600050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Order Info

CapacityMetric

Specifications

Capacity (Metric) 0.45 kg
For Use With Inlet Liquid Catchpots, FL20K Foreline Trap, ITO Inlet Catchpots
Type Activated Alumina
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.