Learn More
Erlab Activated Carbon Filtration At Extraction - Acids And Solvents - Solis 1800
Product Code. 18898080 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Filter
Unit Size:
Each
Product Code. Product Type Unit Size
18898080 Filter Each
1 options
Product Code. 18898080 Supplier Erlab Supplier No. NORT000352/BE
  • ONLINE EXCLUSIVE PRICE
    2405.00 €
    2221.00 € / Pack of 1
    Save 184.00 €
    7% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Filter for your Biological Safety Cabinet.
  • Activated carbon filtration at extraction for your BSC
  • Type of chemicals: Acids and solvents
  • Compatible model: BSC Solis 1800
Specifications
Product Type Filter