Learn More
Erlab Activated Carbon Filtration At Extraction - Solvents - Solis 1200
Product Code. 18553756 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Filter
Unit Size:
Each
Product Code. Product Type Unit Size
18553756 Filter Each
1 options
Product Code. 18553756 Supplier Erlab Supplier No. NORT000350/AS
  • ONLINE EXCLUSIVE PRICE
    1840.00 €
    1697.00 € / Pack of 1
    Save 143.00 €
    7% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Filter for your Biological Safety Cabinet.
  • Activated carbon filtration at extraction for your BSC
  • Type of chemicals: Solvents
  • Compatible model: BSC Solis 1200
Specifications
Product Type Filter