missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT™ A SUPRA

Product Code. 10419640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10419640 250 g Glass bottle 250g
10289873 1 kg Plastic drum 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419640 Supplier Thermo Scientific Acros Supplier No. 134342500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807 | CAS: 7440-44-0 | C | 12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Acidity Passes Test
Melting Point 3652.0°C
Color Black
Loss on Drying 15% max.
Packaging Glass bottle
Quantity 250 g
Merck Index 15, 1807
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5%
Formula Weight 12
Physical Form Powder
Chemical Name or Material Activated charcoal,NORIT* A SUPRA
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.