missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, purum, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Ignition Residue | ≤7.0% |
| Quantity | 1 kg |
| UN Number | NONH for all modes of transport |
| Formula Weight | 12.01g/mol |
| Grade | purum |
| Chemical Name or Material | Activated Charcoal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction