missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Activated Thiol Sepharose™ 4B Medium

Product Code. 11554935 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
15 g
Unit Size:
15g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11554935 15 g 15g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11554935 Supplier Cytiva Supplier No. 17064001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For reversible coupling of proteins to Sepharose 4B via a glutathione spacer arm

  • Gel reacts with heavy metal ions and with alkyl and aryl halides
  • Addition reactions occur with compounds containing C=O, C=C, and N=N bonds
  • Less substituted but more porous than Thiopropyl Sepharose 6B
  • Suitable for covalent chromatography of large molecules such as enzymes and nucleic acids
  • Possible to couple proteins lacking thiol groups to thiol gels using the heterobifunctional reagent SPDP, facilitating use of covalent chromatography

TRUSTED_SUSTAINABILITY

Specifications

Ligand Concentration 1μM Activated Thiol/mL Medium
Matrix 4% Agarose
Particle Size 90 μm
pH 2 to 8
Quantity 15 g
Content And Storage 2°C to 8°C
For Use With (Application) Used for reversible immobilization of molecules containing Thiol groups under mild conditions, optimized for immobilization of large molecules
Material 4% Agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.