missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems ActiveMax™ Human Erythropoietin / EPO Protein, Tag Free

Product Code. 17236200
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17236200 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17236200 Leverantör ACROBiosystems Leverantörsnummer EPOH42141mg

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Human Erythropoietin (EPO) is also known as EP, erythropoetin or erthropoyetin, and is a glycoprotein hormone that controls erythropoiesis, or red blood cell production. EPO is a cytokine for erythrocyte (red blood cell) precursors in the bone marrow. is synthesized by renal peritubular cells in adults, with a small amount being produced in the liver. Regulation is believed to rely on a feed-back mechanism measuring blood oxygenation. Constitutively synthesized transcription factors for EPO, known as hypoxia-inducible factors (HIFs), are hydroxylated and proteosomally digested in the presence of oxygen. It binds to the erythropoietin receptor (EpoR) on the red cell surface and activates a JAK2 cascade. Erythropoietin has its primary effect on red blood cells by promoting red blood cell survival through protecting these cells from apoptosis. It also cooperates with various growth factors involved in the development of precursor red cells. EPO has a range of actions including vasoconstriction-dependent hypertension, stimulating angiogenesis, and inducing proliferation of smooth muscle fibers. It has also been shown that erythropoietin can increase iron absorption by suppressing the hormone hepcidin. Erythropoietin has been shown to interact with the Erythropoietin receptor as its mechanism of action within the body. erythropoietin plays an important role in the brain's response to neuronal injury. EPO is also involved in the wound healing process.

This protein carries no tag. The protein has a calculated MW of 18.4 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifikationer

Accession Number AAH93628.1
Molecular Weight (g/mol) 18.4 kDa
Quantity 1 mg
Source ActiveMax™ Human EPO, Tag Free (EPO-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 28 - Arg 193 (Accession # AAH93628.1).
Immunogen Ala 28 - Arg 193
Storage Requirements -20°C
Gene Alias EPO,EP,MVCD2,Erythropoietin,Erythropoetin,Erthropoyetin,Hematopoietin,Hemopoietin
Common Name Erythropoietin
Conjugate Unconjugated
Species Human
Protein Tag Tag Free
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype ActiveMax™ Cytokines
Purity or Quality Grade >97% as determined by SDS-PAGE.
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.