missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura™ Electro 936 Macropipette Initial Kit

Product Code. 11809233
Change view
Click to view available options
Volume (Metric):
0.5 to 10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11809233 0.5 to 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11809233 Supplier Socorex™ Supplier No. 936.10E
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ergonomic electronic single-channel macropipette initial package. Socorex™ Acura™ Electro 936 Macropipette Initial Kit includes pipette, charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.

  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Interchangeable nozzle protection filter.
  • Adapters for Pasteur pipette and straw tips.
  • Outstanding accuracy and precision.

Specifications

Accuracy <±1.0% - <±0.5%
Battery Type 4.8V NiMH
Certifications/Compliance EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
Model 936
Number of Channels 1
Product Line Acura Electro
Programmability Self-teaching
Type Electronic Macropipette
Volume (Metric) 0.5 to 10 mL
Display Right/Left LCD
Includes Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions.
Increments 0.05mL
Precision <0.5% - <0.15%
Quantity 1 Each
Recharge Time <1.5Hours
Warranty 24 Months
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.