missing translation for 'onlineSavingsMsg'
Learn More

AD-mix-Beta (Technical Grade), TRC

Product Code. 30012886
Change view
Click to view available options
Quantity:
250 mg
10 g
50 g
Unit Size:
10g
250mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012886 10 g 10g
30011489 50 g 50g
30011353 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012886 Supplier Toronto Research Chemicals Supplier No. TRCA20000510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148618-32-0
Molecular Formula C48H54N6O4
Molecular Weight (g/mol) 779
Synonym AD-mix-beta
IUPAC Name 4-[(S)-[(2S,4R,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methoxy]-1-[(R)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methoxy]phthalazine
SMILES CCCCCCC\C=C/CCCCCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30-,31-,32+,43+,44-,45+,46-/m0/s1
Formula Weight 1615.067
Chemical Name or Material AD-mix-beta
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.