Learn More
WTW™ ADA-DIN-BNC Model Adapter
Product Code. 10587451
Change view
Click to view available options
Connector Types:
BNC Plug, DIN Socket
Unit Size:
Each
Product Code. Connector Types Unit Size
10587451 BNC Plug, DIN Socket Each
1 options
Product Code. 10587451 Supplier WTW™ Supplier No. 108509
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Connector Types BNC Plug, DIN Socket
For Use With (Equipment) pH Electrode
Type Adapter
For Use With (Application) Connecting pH Electrodes with BNC Plug to a Meter with DIN Socket