missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carbohydrazide, 97%, Thermo Scientific™

Product Code. 10160842
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160842 1 g Amber Glass Bottle 1g
10275884 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10160842 Supplier Thermo Scientific Maybridge Supplier No. RH01025DA
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 17846-15-0
Molecular Formula C11H18N2O
Molecular Weight (g/mol) 194.278
InChI Key FJXQVRADKYKERU-UHFFFAOYSA-N
Synonym adamantanecarbohydrazide, 1-adamantanecarbohydrazide, adamantane-1-carboxylic acid hydrazide, tricyclo 3.3.1.1∼3,7∼ decane-1-carbohydrazide, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, hydrazide, pubchem21688, hydrazino 1-adamantyl ketone, 3r,5r,7r-adamantane-1-carbohydrazide, z-adamantane-1-carbohydrazonic acid, 3r,5s,7s-adamantane-1-carbohydrazide
PubChem CID 337203
IUPAC Name adamantane-1-carbohydrazide
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)NN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 194.28
Percent Purity 97%
Chemical Name or Material Adamantane-1-carbohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.