missing translation for 'onlineSavingsMsg'
Learn More

Adamantane, 99+%

Product Code. 10602983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602983 5 g Glass bottle 5g
10091510 25 g Glass bottle 25g
10001520 100 g Glass bottle 100g
13458368 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10602983 Supplier Thermo Scientific Acros Supplier No. 146110050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 281-23-2 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 281-23-2
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00074719
InChI Key ORILYTVJVMAKLC-UHFFFAOYSA-N
Synonym tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl
PubChem CID 9238
ChEBI CHEBI:40519
IUPAC Name adamantane
SMILES C1C2CC3CC1CC(C2)C3

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 205.0°C to 210.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Merck Index 15,137
Solubility Information Solubility in water: insoluble. Other solubilities: 8% in light petroleum fract. (20°C),6.7% in diethyl ether-1.7% in ethyl acetate (20°C),9.1% in methylene chloride-0.8% in acetone (20°C),10% in chloroform-12.5% in ccl4 (20°C),0.5%
Formula Weight 136.24
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Adamantane, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.