missing translation for 'onlineSavingsMsg'
Learn More

Adamantane, TRC

Product Code. 30007563
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007563 25 g 25g
30003222 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007563 Supplier Toronto Research Chemicals Supplier No. TRCA20780025G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 281-23-2
Molecular Formula C10 H16
Molecular Weight (g/mol) 136.23
Synonym Adamantane, Tricyclo[3.3.1.13,·]decane, Tricyclo[3.3.1.13,7]decane (9CI, ACI), Adamantane (6CI, 8CI), A 0696, MeSH ID: D000218, NSC 527913
IUPAC Name adamantane
SMILES C1C2CC3CC(CC1C3)C2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H16/c1-7-2-9-4-8(1)5-10(3-7)6-9/h7-10H,1-6H2
Formula Weight 136.1252
Percent Purity >95%
Chemical Name or Material Adamantane
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.