missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Adapalene
GREENER_CHOICE

Product Code. 18085572
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085572 50 mg 50mg
18065052 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085572 Supplier Tocris Bioscience™ Supplier No. 2852/50
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

RARβ and RARγ agonist

Retinoic acid analog that is a RARβ and RARγ agonist (AC50 values are 2.2, 9.3, 22 and > 1000 nM for RARβ, RARγ, RARα and RXRα receptors respectively). Inhibits proliferation and induces apoptosis in colorectal cancer cell in vitro. Displays comedolytic activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106685-40-9
Molecular Formula C28H28O3

Specifications

Quantity 50 mg
Formula Weight 412.52
Purity >98%
Molecular Weight (g/mol) 412.529
Chemical Name or Material Adapalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.