Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Adapter for Rotors designed for use with Eppendorf Centrifuges
Brand:  Eppendorf™ 5427731008
Code : A0
Additional Details : Weight : 0.04700kg
 View more versions of this product
Product Code. 10628004
  • POA / Pack of 2
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
Specifications
Specifications
Small Rotor Bore Adapter
1 x 2.6 to 7 mL Tube
Small rotor bore
Round Bottom
Rotor F-35-6-30
2/Pk.