missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapter for Rotor T-60-11

Designed for use with Eppendorf Centrifuges

Brand:  Eppendorf™ 5804732006

218.54 EUR valid until 2023-12-31
Use promo code "20333" to get your promotional price.



Code : 99

Additional Details : Weight : 1.05000kg

 View more versions of this product

Product Code. 10053361

  • 230.52 € / Pack of 6
Estimated Shipment: 14-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High capacity for 60 × 1.5/2.0mL tubes
  • Max. speed: 16,400 × g (14,000 rpm)
  • Horizontal centrifugation for pellet formation at tube bottom
  • Ideal for low sample quantities
Specifications

Specifications

Adapter
Rotor T-60-11
20 Reaction
Low sample quantities
0.4 mL
6 mm
120
6 Pack
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers