Learn More
Eppendorf™ Adapter for HPLC Vials
Product Code. 13236619 Shop All Eppendorf Products
Change view
Click to view available options
No. per Pack:
1/Pk.
Unit Size:
Pack of 10
Product Code. No. per Pack Unit Size
13236619 1/Pk. Pack of 10
1 options
Product Code. 13236619 Supplier Eppendorf™ Supplier No. 5820770007
  • ONLINE EXCLUSIVE PRICE
    45.83 €
    41.50 € / Pack of 4
    Save 4.33 €
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for use with Eppendorf Rotor FA-45-20-17
  • Adapter for 1 HPLC Vial
  • For Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
  • Available in packs of 4
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
For Use With (Equipment) Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
For Use With (Application) HPLC
No. per Pack 1/Pk.