missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Adapter Vials 15 x 42 mm

Product Code. 31777384 Shop All Berthold Technologies Products
Change view
Click to view available options
Includes:
Adapters (10 Pc.)
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
31777384 Adapters (10 Pc.) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777384 Supplier Berthold Technologies Supplier No. 32737
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Junior portable tube luminometers.

  • Prevent tubes from moving during measurement
  • Protect the sample chamber from contamination
  • Contain reagent spills within the adapter
  • Allow quick replacement of the adapter

Specifications

Color Transparent
For Use With (Equipment) Junior LB 9509 Portable Luminometer
Height (Metric) 42 mm
Width (Metric) 15 mm
Includes Adapters (10 Pc.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.