Learn More
WTW™ Adapter for WTW™ FDO 925
Product Code. 15229094
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material Unit Size
15229094 Stainless Steel Each
1 options
Product Code. 15229094 Supplier WTW™ Supplier No. 201312
  • ONLINE EXCLUSIVE PRICE
    53.30 €
    51.00 € / Pack of 1
    Save 2.30 €
    4% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
For Use With (Equipment) Mounting a RZ 300 Magnetic Stirrer Paddle for BOD Measurement on a FDO™ 925
Type Adapter
Material Stainless Steel