missing translation for 'onlineSavingsMsg'
Learn More

Carl Zeiss™ Adapters

Product Code. 12574346
Change view
Click to view available options
Type:
Adapter 60N - T2, 1.0X
DSLR P95-T2 1.6x Adapter
Preparation Piece 60-44
Video Adapter 44, 0.5 in., 0.5X
Video Adapter 60 C, 0.5 in., 0.5X
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12574346 Adapter 60N - T2, 1.0X Each
10738683 Preparation Piece 60-44 Each
11810903 Video Adapter 44, 0.5 in., 0.5X Each
11866472 Video Adapter 60 C, 0.5 in., 0.5X Each
11855143 DSLR P95-T2 1.6x Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12574346 Supplier Carl Zeiss™ Supplier No. 4261030000000
  • ONLINE EXCLUSIVE PRICE
    71.00 €
    63.03 € / Pack of 1
    Save 7.97 €
    11% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.