missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters For Fixed-Angle Rotor

Add flexibility to Eppendorf Centrifuges with these rotor adapters

Brand:  Eppendorf™ 5804770005

Code : 99

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 10043061

  • 137.05 € / Pack of 2
Estimated Shipment: 25-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Adapters
  • For Eppendorf™ Fixed Angle Rotors
Specifications

Specifications

Adapter
1.5 to 2 mL
11 mm
2 Pack
Eppendorf
Fixed angle rotor
24
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers