missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Adavosertib

Product Code. 18777586
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777586 10 mg 10mg
18758086 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18777586 Supplier R&D Systems™ Supplier No. 7589/10
  • ONLINE EXCLUSIVE PRICE
    250.66 €
    225.75 € / 10mg
    Save 24.91 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective Wee1 inhibitor

Adavosertib is a potent and selective Wee1 inhibitor (IC50 = 5.2 nM). In a panel of 223 serine/threonine or tyrosine kinases, Adavosertib is highly selective for Wee 1 over all except 8 kinases. In particular Adavosertib shows >100-fold selectivity over human Myt1. Adavosertib blocks CDC2Y15 phosphorylation and Gemcitabine (Cat. No. 3259)-induced DNA damage checkpoint, leading to premature mitotic entry and apoptosis in p53-deficient tumor cell lines. Adavosertib also potentiates the cytotoxic effects of Pemetrexed (Cat. No. 6185), Doxorubicin (Cat. No. 2252), Camptothecin (Cat. No. 1100) and Mitomycin C (Cat. No. 3258) in vitro. Adavosertib synergizes with gemcitabine to achieve tumor regressions, selectively in p53-deficient pancreatic cancer xenografts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dihydro-1-[6-(1-hydroxy-1-methylethyl)-2-pyridinyl]-6-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-2-(2-propen-1-yl)-3 H-pyrazolo[3,4-d]pyrimidin-3-one
CAS 955365-80-7
Quantity 10 mg
Product Line Tocris
Target Checkpoint Control Kinase Inhibitors
Molecular Formula C27H32N8O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.