missing translation for 'onlineSavingsMsg'
Learn More

Adavosertib, MedChemExpress

Product Code. 16444338
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444338 10 mg 10mg
16417057 5 mg 5mg
16400667 50 mg 50mg
16415256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444338 Supplier Medchem Express Supplier No. HY10993
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Adavosertib (AZD-1775; MK-1775) is a potent Wee1 inhibitor with an IC50 of 5.2 nM.

Adavosertib (AZD-1775

Specifications

Chemical Name or Material Adavosertib
CAS 955365-80-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H32N8O2
Quantity 10 mg
Synonym AZD1775 MK-1775
Solubility Information DMSO : 125 mg/mL (249.70 mM; Need ultrasonic)
SMILES O=C1N(N(C2=NC(NC3=CC=C(C=C3)N4CCN(CC4)C)=NC=C21)C5=NC(C(O)(C)C)=CC=C5)CC=C
Molecular Weight (g/mol) 500.6
Formula Weight 500.6
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.