missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ADCF-MAb Media

Product Code. 10094723
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 1000 mL
Unit Size:
1000mL
500mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10094723 6 x 1000 mL Pack of 6
10788134 500 mL 500mL
10397042 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10094723 Supplier Cytiva Supplier No. SH30349.LS
  • ONLINE EXCLUSIVE PRICE
    456.00 €
    315.49 € / Pack of 6
    Save 140.51 €
    30% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.

  • ADCF
  • Metabolically designed for high cell yield and monoclonal antibody production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Contains L-glutamine and Poloxamer 188
  • Formulations without L-glutamine support the GS gene expression system
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type ADCF-mAb Liquid Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Poloxamer 188, HEPES, Sodium Bicarbonate
Without Additives No Protein, No Phenol Red
Quantity 6 x 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description with L-Glutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.