missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Additional Perforated Stainless Steel Shelf for UN160

Product Code. 17579845 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN160 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17579845 UN160 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17579845 Supplier Memmert™ Supplier No. UN160B00325
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Shelves with a perforated stainless steel construction for ovens and incubators.

  • Perforated Stainless Steel Shelf
  • Accessory for Memmert ovens and incubators

Specifications

For Use With (Equipment) UN160 Oven
Type Perforated Shelf
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.