missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Additional Perforated Stainless Steel Shelf for UN30

Product Code. 17590056 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17590056 UN30 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17590056 Supplier Memmert™ Supplier No. UN30B29727
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Shelves with a perforated stainless steel construction for ovens and incubators.

  • Perforated Stainless Steel Shelf
  • Accessory for Memmert ovens and incubators

Specifications

For Use With (Equipment) UN30 Oven
Type Perforated Shelf
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.