Learn More
Memmert™ Additional Stainless Steel Grid, electro-polished, For Icp450
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Models 450 and 750 Ovens
Unit Size:
Each
Description
- For use with compressor-cooled incubator models 450 and 750
- Grid pattern allows for airflow
- Manufactured from electro-polished stainless steel
Specifications
Specifications
| For Use With (Equipment) | Models 450 and 750 Ovens |
| Material | Stainless Steel |
| Type | Grid |