missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine, 99%

Product Code. 10444595 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10444595 500 g Glass bottle 500g
10716181 25 g Glass bottle 25g
10746341 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444595 Supplier Thermo Scientific Chemicals Supplier No. 147445000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73-24-5 | C5H5N5 | 135.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Absorbance 0.74 to 0.78 (A250/A260), 0.36 to 0.4 (A280/A260)
CAS Min % 99.5
CAS Max % 100.0
Melting Point 360°C to 365°C
Color Colorless to Yellow
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 g
Beilstein 26, 420
Merck Index 15, 140
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in acids and alkalies,slightly soluble in ethanol,insoluble in chloroform and ether,25g/l in boiling water
Sublimation Point 220 °C
Spectrophotometry 98.0 to 102.0% (lambda max. 262nm, pH 1), molar absorptivity 13100 +/- 2%, on dry basis
Formula Weight 135.13
Percent Purity 99%
Physical Form Liquid or Solid
Chemical Name or Material Adenine
Show More Show Less
compliance-icons
Product Identifier
  • Adenine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.