missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine, 99%

Product Code. 11412717
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
Unit Size:
250g
25g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412717 500 g 500g
11402717 25 g 25g
11304619 50 g 50g
11314619 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11412717 Supplier Thermo Scientific Chemicals Supplier No. A14906.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73-24-5 | C5H5N5 | 135.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water (3 g/L) at 20°C.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Melting Point >360°C (decomposition)
Assay Percent Range 99%
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 5777
Merck Index 14,152
Solubility Information Soluble in hot water,methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.
Formula Weight 135.13
Percent Purity 99%
Chemical Name or Material Adenine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.