missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hemisulfate Dihydrate MP Biomedicals

Product Code. 11348519 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348519 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11348519 Supplier MP Biomedicals Supplier No. 0210019525
  • ONLINE EXCLUSIVE PRICE
    129.00 €
    121.80 € / 25g
    Save 7.20 €
    5% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tekniske data

CAS 6509-19-9
Assay Percent Range 99%
Molecular Formula C10H16N10O6S
MDL Number MFCD00150149
Quantity 25 g
Synonym 9h-purin-6-amine hemisulfate dihydrate, adenine hemisulfate dihydrate, 6-aminopurine sulfate dihydrate, bis ade sulfuric acid dihydrate, sulfuric acid bis vitamin b4 dihydrate, bis adenine sulfuric acid dihydrate, sulfuric acid-7h-purin-6-amine-water 1/2/2
InChI Key MKPCNMXYTMQZBE-UHFFFAOYSA-N
SMILES O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
IUPAC Name bis(7H-purin-6-amine) sulfuric acid dihydrate
Molecular Weight (g/mol) 404.36
PubChem CID 44134512
Formula Weight 404.4
Chemical Name or Material Adenine Hemisulfate, Dihydrate
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.