missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine hydrochloride, 98+%, cont. up to ca 5% water

Product Code. 11428257
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428257 100 g 100g
11408257 5 g 5g
11418257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428257 Supplier Thermo Scientific Chemicals Supplier No. A17622.22
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2922-28-3 | C5H6ClN5 | 171.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2922-28-3
Molecular Formula C5H6ClN5
Molecular Weight (g/mol) 171.59
MDL Number MFCD00038990
InChI Key UQVDQSWZQXDUJB-UHFFFAOYSA-N
Synonym adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
PubChem CID 76219
IUPAC Name 7H-purin-6-amine;hydrochloride
SMILES Cl.NC1=C2NC=NC2=NC=N1

Specifications

Melting Point ∼290°C (decomposition)
Quantity 100 g
Beilstein 4009585
Formula Weight 171.59
Percent Purity ≥98%
Assay cont. up to ≈5% water
Chemical Name or Material Adenine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.