missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine sulfate, 98+%

Product Code. p-7023420
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458796 10 g 10g
11468796 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458796 Supplier Thermo Scientific Chemicals Supplier No. A16964.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol

It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.

Solubility
Soluble in water (4 mg/ml), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and HCl (10 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name 7H-purin-6-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Melting Point ∽210°C (decomposition)
Assay Percent Range >98%
Quantity 10 g
Beilstein 3820263
Sensitivity Hygroscopic
Merck Index 14,152
Solubility Information Soluble in water (4mg/ml),ethanol (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),and HCl (10mg/ml).
Formula Weight 368.34
Percent Purity ≥98%
Chemical Name or Material Adenine sulfate
compliance-icons
Product Identifier
  • Adenine sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Purin-6-amine, sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.