missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine sulfate, 98%, synthetic

Product Code. 10636562 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636562 25 g Plastic bottle 25g
10646562 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10636562 Supplier Thermo Scientific Chemicals Supplier No. 163630250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-30-2
Molecular Formula C10H12N10O4S
Molecular Weight (g/mol) 368.33
MDL Number MFCD00213655
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
PubChem CID 9449
IUPAC Name 7H-purin-6-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Absorbance 0.74 to 0.78 (A250/A260), 0.34 to 0.38 (A280/A260)
CAS Min % 97.5
CAS Max % 100.0
Melting Point 285.0°C
Color White to Yellow
Loss on Drying 2% max. (1 g, 105°C, 5 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 26, V,16, 100
Solubility Information Solubility in water: soluble
Spectrophotometry 97.5% min. (lambda max. 262nm,pH 1) molar absorptivity 13100+/-2%,on dry basis
Transmittance 95% min. (c=0.5, H2O, 430nm) 1cm cell
Formula Weight 184.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenine sulfate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.