missing translation for 'onlineSavingsMsg'
Learn More

Adenine Sulfate, MP Biomedicals

Product Code. 11397769 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397769 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11397769 Supplier MP Biomedicals Supplier No. 114014522
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 321-30-2
Molecular Formula C10H12N10O4S
MDL Number MFCD00213655
Quantity 10 g
Synonym adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
IUPAC Name bis(7H-purin-6-amine); sulfuric acid
Molecular Weight (g/mol) 368.33
PubChem CID 9449
Physical Form Powder
Chemical Name or Material ADENINE SULFATE
Show More Show Less
compliance-icons
Product Identifier
  • Adenine Hemisulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.