missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Adenine
GREENER_CHOICE

Product Code. 18074892
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074892 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18074892 Supplier Tocris Bioscience™ Supplier No. 4202/50
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Adenine receptor agonist

High affinity adenine receptor agonist (Ki = 18 nM at rat adenine receptor). Inhibits forskolin-stimulated cAMP formation in CHO cells transfected with the adenine receptor; also stimulates GTPγS binding (pEC50 values are 8.54 and 7.21 respectively).
TRUSTED_SUSTAINABILITY

Specifications

CAS 2922-28-3
Quantity 50 mg
Molecular Formula C5H6ClN5
Formula Weight 171.59
Purity >99%
Molecular Weight (g/mol) 171.59
Chemical Name or Material Adenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.