missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-3',5'-cyclic monophosphate, 98%

Codice prodotto. 15463619
Click to view available options
Quantity:
1 g
5 g
Dimensione della confezione:
1g
5g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 15463619

Marca: Thermo Scientific Chemicals J60936.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 60-92-4 | C10H12N5O6P | 329.21 g/mol

cAMP is used for intracellular signal transduction in many different organisms, conveying the cAMP-dependent pathway. Restoration of several morphological characteristics of normal fibroblasts in sarcoma cells when treated with adenosine-3?: 5?-cyclic monophosphate. In the presence of adenosine 3?:5?-cyclic monophosphate, protein kinase I dissociated into two subunits: a subunit binding adenosine 3?:5?-cyclic monophosphate, and a catalytic subunit. Stimulation of calcium uptake in platelet membrane vesicles by adenosine 3?,5?-cyclic monophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cAMP is used for intracellular signal transduction in many different organisms, conveying the cAMP-dependent pathway. Restoration of several morphological characteristics of normal fibroblasts in sarcoma cells when treated with adenosine-3′: 5′-cyclic monophosphate. In the presence of adenosine 3′:5′-cyclic monophosphate, protein kinase I dissociated into two subunits: a subunit binding adenosine 3′:5′-cyclic monophosphate, and a catalytic subunit. Stimulation of calcium uptake in platelet membrane vesicles by adenosine 3′,5′-cyclic monophosphate.

Solubility
Solube in water: 10 mg/mL pH of aqueous solution is approx. 3.0.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 60-92-4
Molecular Formula C10H12N5O6P
Molecular Weight (g/mol) 329.21
MDL Number MFCD00005845
InChI Key IVOMOUWHDPKRLL-YPLCUDRINA-N
Synonym camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
PubChem CID 6076
ChEBI CHEBI:17489
IUPAC Name (4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
SMILES NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifica

Melting Point 260°C (decomposition)
Color White
Quantity 1 g
Beilstein 52645
Merck Index 14,2708
Solubility Information Solube in water: 10mg/mL pH of aqueous solution is approx. 3.0.
Formula Weight 329.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-3',5'-cyclic monophosphate

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato