missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-3'-monophosphate, TRC

Product Code. 30240976
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
Unit Size:
200mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240976 200 mg 200mg
30240549 25 mg 25mg
30240548 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240976 Supplier Toronto Research Chemicals Supplier No. TRCA281840200MG
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-21-9
Molecular Formula C10H14N5O7P, C10 H14 N5 O7 P

Specifications

Format Neat
Molecular Weight (g/mol) 347.2212, 347.22
MDL Number MFCD00005746
InChI Key LNQVTSROQXJCDD-KQYNXXCUSA-N
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](OP(O)(O)=O)[C@H]3O)C2=NC=N1
IUPAC Name {[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}phosphonic acid
Shipping Condition Room Temperature
Quantity 200 mg
Chemical Name or Material Adenosine-3'-monophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.