missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-diphosphate trilithium salt, 98%

Product Code. 15446429
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446429 1 g 1g
15456429 5 g 5g
15466429 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15446429 Supplier Thermo Scientific Chemicals Supplier No. J64497.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 31008-64-7 | C10H12Li3N5O10P2 | 445.00 g/mol

It is used as a salt formulation of the essential metabolic compound adenosine diphosphate (ADP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a salt formulation of the essential metabolic compound adenosine diphosphate (ADP).

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31008-64-7
Molecular Formula C10H12Li3N5O10P2
Molecular Weight (g/mol) 445.00
MDL Number MFCD00065469
InChI Key LZGPPAHUZSOGHJ-DJXXCMMGNA-K
Synonym adenosine-5'-diphosphate trilithium salt, adenosine 5'-trihydrogen diphosphate , trilithium salt, adp-li3, trilithium 1+ adenosine 5'-diphosphate, adenosine 5'-diphosphate trilithium salt, adenosine 5'-diphosphate,trilithium salt, adenosine-5'-diphosphate, trilithium salt, trilithium 1+ ion adenosine 5'-diphosphate, adenosine 5'-diphosphoric acid trilithium salt, adenosine-5'-diphosphate,trilithium salt
PubChem CID 56841973
IUPAC Name trilithium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate
SMILES [Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 445
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenosine-5'-diphosphate trilithium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.