missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-monophosphate disodium salt

Product Code. 15474199
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474199 5 g 5g
15484199 25 g 25g
15494199 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15474199 Supplier Thermo Scientific Alfa Aesar Supplier No. J61643.06
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4578-31-8 | C10H12N5Na2O7P | 391.19 g/mol

Adenosine-5'-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5'-nucleotidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5′-nucleotidase.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Air and moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4578-31-8
Molecular Formula C10H12N5Na2O7P
Molecular Weight (g/mol) 391.19
MDL Number MFCD00065023
InChI Key QGXLVXZRPRRCRP-IDIVVRGQSA-L
Synonym adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt
PubChem CID 20712
ChEBI CHEBI:81690
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifikationer

Color White
Quantity 5 g
Beilstein 3586732
Merck Index 14,158
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
Formula Weight 391.2
Physical Form Powder
Chemical Name or Material Adenosine-5'-monophosphate disodium salt

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.