missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-triphosphate disodium salt hydrate Thermo Scientific Chemicals

Product Code. 15895048 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895048 5 g Plastic bottle 5g
15805058 10 g Plastic bottle 10g
15815058 25 g Plastic bottle 25g
15825058 100 g Plastic bottle 100g
15835058 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15895048 Supplier Thermo Scientific Supplier No. J10585.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34369-07-8 | C10H20N5Na2O16P3 | 605.19 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34369-07-8
Molecular Formula C10H20N5Na2O16P3
Molecular Weight (g/mol) 605.19
MDL Number MFCD00150754
InChI Key MWEQTWJABOLLOS-MVHBFMHHNA-L
Synonym adenosine 5 '-triphosphate disodium salt trihydrate, adenosine 5 '-triphosphate disodium salt, adenosine 5 '-triphosphate disodium salt trihydrate, disodium salt
PubChem CID 134129625
IUPAC Name ({[(2 R,3 S,4 R,5 R)-5-(6-amino-9 H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
SMILES O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 551.14 (Anhydrous)
Packaging Plastic bottle
Chemical Name or Material Adenosine-5 '-Triphosphate Disodium Salt Trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.