missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine, 99%

Product Code. 11439692
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439692 10 g 10g
11449692 50 g 50g
11459692 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439692 Supplier Thermo Scientific Chemicals Supplier No. A10781.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol

Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.

Solubility
Soluble in water, ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Melting Point 234°C to 237°C
Assay Percent Range 99%
Odor Odorless
Quantity 10 g
Beilstein 93029
Merck Index 14,153
Solubility Information Soluble in water,ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.
Optical Rotation −70° (c=2 in 5% NaOH)
Formula Weight 267.25
Percent Purity 99%
Chemical Name or Material Adenosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.