missing translation for 'onlineSavingsMsg'
Learn More

Adenosine, 99.8%, MP Biomedicals™

Product Code. 11318529 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318529 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11318529 Supplier MP Biomedicals Supplier No. 0210019991
  • ONLINE EXCLUSIVE PRICE
    1925.00 €
    1824.90 € / 1kg
    Save 100.10 €
    5% Off
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A nucleotide resulting from the union of one molecule of sugar -- d-ribose -- with a base, l-adenine

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Density 2.09g/cm3
Assay Percent Range 99.80%
Quantity 1 kg
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Formula Weight 267.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.